2-Methyl-5-(tributylstannyl)furan ≥97%
Supplier: Matrix Scientific
069555-500MG
069555-1G
069555-5G
101940-778EA
263.08
USD
101940-778
101940-780
101940-782
2-Methyl-5-(tributylstannyl)furan ≥97%
2-Methyl-5-(tributylstannyl)furan
Hazard: Irritant
Formula:
C₁₇H₃₂OSn |
MDL Number:
MFCD01319081 CAS Number: 118486-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...