3,5-Dimethyl-4-(tributylstannyl)isoxazole ≥95%

Supplier: Matrix Scientific

069554-500MG 069554-1G 069554-5G
101940-772EA 263.08 USD
101940-772 101940-774 101940-776
3,5-Dimethyl-4-(tributylstannyl)isoxazole ≥95%
3,5-Dimethyl-4-(tributylstannyl)isoxazole

Hazard: Irritant

Formula: C₁₇H₃₃NOSn
MDL Number: MFCD01318973
CAS Number: 136295-80-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR