2-Fluoro-5-methyl-3-(tributylstannyl)pyridine
Supplier: Matrix Scientific
069553-500MG
069553-1G
069553-5G
101940-766EA
349.23
USD
101940-766
101940-768
101940-770
2-Fluoro-5-methyl-3-(tributylstannyl)pyridine
2-Fluoro-5-methyl-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₂FNSn |
MDL Number:
MFCD09025799 CAS Number: 1025745-87-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...