2-Fluoro-3-methyl-5-(tributylstannyl)pyridine

Supplier: Matrix Scientific

069552-500MG 069552-1G 069552-5G
101940-760EA 349.23 USD
101940-760 101940-762 101940-764
2-Fluoro-3-methyl-5-(tributylstannyl)pyridine
2-Fluoro-3-methyl-5-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₂FNSn
MDL Number: MFCD09025798
CAS Number: 405556-98-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR