3,5-Difluoro-4-(tributylstannyl)pyridine ≥90%

Supplier: Matrix Scientific

069551-1G 069551-5G
101940-756EA 349.23 USD
101940-756 101940-758
3,5-Difluoro-4-(tributylstannyl)pyridine ≥90%
3,5-Difluoro-4-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₂₉F₂NSn
MDL Number: MFCD09025762
CAS Number: 765916-82-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR