3-Formyl-5-(tributylstannyl)pyridine dimethylacetal ≥98%

Supplier: Matrix Scientific

069550-500MG 069550-1G 069550-5G
101940-750EA 466.15 USD
101940-750 101940-752 101940-754
3-Formyl-5-(tributylstannyl)pyridine dimethylacetal ≥98%
3-Formyl-5-(tributylstannyl)pyridine dimethylacetal

Hazard: Irritant

Formula: C₂₂H₄₁NO₃Sn
MDL Number: MFCD10699129
CAS Number: 1264193-67-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR