3-Formyl-2-(tributylstannyl)furan

Supplier: Matrix Scientific

069549-500MG 069549-1G 069549-5G
101940-744EA 263.08 USD
101940-744 101940-746 101940-748
3-Formyl-2-(tributylstannyl)furan
3-Formyl-2-(tributylstannyl)furan

Hazard: Irritant

Formula: C₁₇H₃₀O₂Sn
MDL Number: MFCD09025794
CAS Number: 130056-68-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR