2-Methoxy-5-(tributylstannyl)thiazole ≥95%

Supplier: Matrix Scientific

069548-500MG 069548-1G 069548-5G
101940-738EA 349.23 USD
101940-738 101940-740 101940-742
2-Methoxy-5-(tributylstannyl)thiazole ≥95%
2-Methoxy-5-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₃₁NOSSn
MDL Number: MFCD09025806
CAS Number: 1025744-42-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR