2-Ethoxy-5-(tributylstannyl)thiazole ≥97%

Supplier: Matrix Scientific

069547-500MG 069547-1G 069547-5G
101940-732EA 284.62 USD
101940-732 101940-734 101940-736
2-Ethoxy-5-(tributylstannyl)thiazole ≥97%
2-Ethoxy-5-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₇H₃₃NOSSn
MDL Number: MFCD09025788
CAS Number: 446285-61-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR