2-(Methylthio)-5-(tributylstannyl)pyridine ≥95%
Supplier: Matrix Scientific
069546-500MG
069546-1G
069546-5G
101940-726EA
349.23
USD
101940-726
101940-728
101940-730
2-(Methylthio)-5-(tributylstannyl)pyridine ≥95%
2-(Methylthio)-5-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NSSn |
MDL Number:
MFCD09025805 CAS Number: 611168-64-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...