2-Methoxy-6-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

069544-500MG 069544-1G 069544-5G
101940-714EA 153.85 USD
101940-714 101940-716 101940-718
2-Methoxy-6-(tributylstannyl)pyridine ≥96%
2-Methoxy-6-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₃NOSn
MDL Number: MFCD09025803
CAS Number: 164014-94-2
UN: 2788
ADR: 6.1,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR