4-Bromo-2-(tributylstannyl)thiazole ≥97%

Supplier: Matrix Scientific

069542-500MG 069542-1G 069542-5G
101940-702EA 304.75 USD
101940-702 101940-704 101940-706
4-Bromo-2-(tributylstannyl)thiazole ≥97%
4-Bromo-2-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₅H₂₈BrNSSn
MDL Number: MFCD09025792
CAS Number: 173978-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR