2-Acetamido-5-bromothiazole ≥98%

Supplier: Matrix Scientific

Synonyms: N-(5-bromothiazol-2-yl)acetamide

069541-1G 069541-5G 069541-25G
101940-696EA 263.08 USD
101940-696 101940-698 101940-700
2-Acetamido-5-bromothiazole ≥98%
2-Acetamido-5-bromothiazole

Hazard: Irritant

Formula: C₅H₅BrN₂OS
MW: 221.0800 g/mol
MDL Number: MFCD00017336
CAS Number: 7336-54-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR