4-Chloro-2-(methylthio)pyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2-methylthiopyrimidine

069539-25G 069539-100G
101940-692EA 64.62 USD
101940-692 102663-854
4-Chloro-2-(methylthio)pyrimidine ≥98%
4-Chloro-2-(methylthio)pyrimidine

Hazard: Corrosive/Moisture Sensitive/Stench

Formula: C₅H₅ClN₂S
MW: 160.63 g/mol
Boiling Pt: 118-120°C/9mm
Density: 1.38
MDL Number: MFCD00006083
CAS Number: 49844-90-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR