2,6-Dimethoxy-3-pyridineboronic acid ≥95%
Supplier: Matrix Scientific
Synonyms:
2,6-Dimethoxypyridine-3-boronic acid, 2,6-Dimethoxypyridin-3-ylboronic acid
069535-1G
069535-5G
069535-25G
101940-668EA
233.85
USD
101940-668
101940-670
101940-672
2,6-Dimethoxy-3-pyridineboronic acid ≥95%
2,6-Dimethoxy-3-pyridineboronic acid
Hazard: Irritant
Formula:
C₇H₁₀BNO₄ |
MDL Number:
MFCD03788239 CAS Number: 221006-70-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...