3-Bromo-5-(2-hydroxyethyl)isoxazole ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(3-Bromoisoxazol-5-yl)ethanol

069532-500MG 069532-1G 069532-5G
101940-652EA 356.92 USD
101940-652 101940-654 101940-656
3-Bromo-5-(2-hydroxyethyl)isoxazole ≥95%
3-Bromo-5-(2-hydroxyethyl)isoxazole

Hazard: Irritant

Formula: C₅H₆BrNO₂
MW: 192.02 g/mol
MDL Number: MFCD09025786
CAS Number: 105175-00-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR