Ethyl-2-isopropylthiazole-4-carboxylate ≥96%
Supplier: Matrix Scientific
Synonyms:
Ethyl 2-isopropylthiazole-4-carboxylate
069531-1G
069531-5G
069531-25G
101940-646EA
192.31
USD
101940-646
101940-648
101940-650
Ethyl-2-isopropylthiazole-4-carboxylate ≥96%
Ethyl-2-isopropylthiazole-4-carboxylate
Hazard: Irritant
Formula:
C₉H₁₃NO₂S MW: 199.27 g/mol |
MDL Number:
MFCD09025787 CAS Number: 133047-44-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...