2-Formyl-4-(tributylstannyl)thiazole ≥95%

Supplier: Matrix Scientific

069528-500MG 069528-1G 069528-5G
101940-630EA 698.46 USD
101940-630 101940-632 101940-634
2-Formyl-4-(tributylstannyl)thiazole ≥95%
2-Formyl-4-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₆H₂₉NOSSn
MDL Number: MFCD09025791
CAS Number: 231278-14-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR