5-Bromo-2-morpholin-1-ylpyrimidine ≥98%

Supplier: Matrix Scientific

Synonyms: 5-Bromo-2-(4-morpholino)pyrimidine, 4-(5-Bromopyrimidin-2-yl)morpholine

069526-1G 069526-5G 069526-25G
101940-618EA 44.62 USD
101940-618 101940-620 101940-622
5-Bromo-2-morpholin-1-ylpyrimidine ≥98%
5-Bromo-2-morpholin-1-ylpyrimidine

Hazard: Irritant

Formula: C₈H₁₀BrN₃O
MW: 244.1 g/mol
MDL Number: MFCD00483251
CAS Number: 84539-22-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR