2-Chloro-6-methoxypyrazine ≥98%

Supplier: Matrix Scientific

069525-1G 069525-5G 069525-25G
101940-612EA 58.46 USD
101940-612 101940-614 101940-616
2-Chloro-6-methoxypyrazine ≥98%
2-Chloro-6-methoxypyrazine

Hazard: Irritant

Formula: C₅H₅ClN₂O
MW: 144.5600 g/mol
MDL Number: MFCD06410937
CAS Number: 33332-30-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR