4-(Aminomethyl)-2-isopropylthiazole dihydrochloride ≥96%

Supplier: Matrix Scientific

069524-500MG 069524-1G 069524-5G
101940-606EA 349.23 USD
101940-606 101940-608 101940-610
4-(Aminomethyl)-2-isopropylthiazole dihydrochloride ≥96%
4-(Aminomethyl)-2-isopropylthiazole dihydrochloride

Hazard: Irritant

Formula: C₇H₁₄Cl₂N₂S
MW: 229.17 g/mol
MDL Number: MFCD08275729
CAS Number: 1171981-10-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR