4-Benzyloxy-1-tert-butoxycarbonylindole ≥98%

Supplier: Matrix Scientific

069522-1G 069522-5G
101940-596EA 233.85 USD
101940-596 101940-598
4-Benzyloxy-1-tert-butoxycarbonylindole ≥98%
4-Benzyloxy-1-tert-butoxycarbonylindole

Hazard: Irritant

Formula: C₂₀H₂₁NO₃
MDL Number: MFCD08275732
CAS Number: 903131-59-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR