N-(tert-Butoxycarbonyl)propargylamine ≥98%

Supplier: Matrix Scientific

Synonyms: N-Boc-propargylamine, 2-Propynylcarbamic Acid tert-Butyl Ester, tert-Butyl 2-Propynylcarbamate

069519-5G 069519-25G
101940-580EA 61.54 USD
101940-580 101940-582
N-(tert-Butoxycarbonyl)propargylamine ≥98%
N-(tert-Butoxycarbonyl)propargylamine

Hazard: Irritant

Formula: C₈H₁₃NO₂
MDL Number: MFCD07367245
CAS Number: 92136-39-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR