2-Methoxy-6-(tributylstannyl)pyrazine ≥95%

Supplier: Matrix Scientific

069517-1G 069517-5G 069517-25G
101940-566EA 756.92 USD
101940-566 101940-568 101940-570
2-Methoxy-6-(tributylstannyl)pyrazine ≥95%
2-Methoxy-6-(tributylstannyl)pyrazine

Hazard: Irritant

Formula: C₁₇H₃₂N₂OSn
MW: 399.15
MDL Number: MFCD08275735
CAS Number: 1105511-66-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR