3-(Ethylsulfonyl)phenylboronic acid ≥98%

Supplier: Matrix Scientific

069516-1G 069516-5G
101940-562EA 960 USD
101940-562 101940-564
3-(Ethylsulfonyl)phenylboronic acid ≥98%
3-(Ethylsulphonyl)phenylboronic acid

Hazard: Irritant

Formula: C₈H₁₁BO₄S
MDL Number: MFCD03095257
CAS Number: 845870-47-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR