4-(Cyclopropylaminocarbonyl)phenylboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
4-(Cyclopropylcarbamoyl)benzeneboronic acid, 4-(Cyclopropylaminocarbonyl)phenylboronic acid
069515-1G
069515-5G
101940-558EA
610.77
USD
101940-558
101940-560
4-(Cyclopropylaminocarbonyl)phenylboronic acid ≥97%
4-(Cyclopropylcarbamoyl)phenylboronic acid
Hazard: Irritant
Formula:
C₁₀H₁₂BNO₃ |
MDL Number:
MFCD03788419 CAS Number: 515140-26-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...