4-(Cyclopentylcarbamoyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Cyclopentylaminocarbonyl)phenylboronic acid

069514-1G 069514-5G 069514-25G
101940-552EA 175.38 USD
101940-552 101940-554 101940-556
4-(Cyclopentylcarbamoyl)phenylboronic acid ≥97%
4-(Cyclopentylcarbamoyl)phenylboronic acid

Hazard: Irritant

Formula: C₁₂H₁₆BNO₃
MDL Number: MFCD03788418
CAS Number: 850568-15-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR