2-Fluoro-5-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

069512-500MG 069512-1G 069512-5G
101940-542EA 233.85 USD
101940-542 101940-544 101940-546
2-Fluoro-5-(tributylstannyl)pyridine ≥96%
2-Fluoro-5-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀FNSn
MDL Number: MFCD07787406
CAS Number: 1025687-41-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR