2-Fluoro-4-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

069511-500MG 069511-1G 069511-5G
101940-536EA 524.62 USD
101940-536 101940-538 101940-540
2-Fluoro-4-(tributylstannyl)pyridine ≥96%
2-Fluoro-4-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₇H₃₀FNSn
CAS Number: 457061-31-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR