4-Methyl-2-(tributylstannyl)pyridine ≥93%
Supplier: Matrix Scientific
069509-500MG
069509-1G
069509-5G
101940-524EA
466.15
USD
101940-524
101940-526
101940-528
4-Methyl-2-(tributylstannyl)pyridine ≥93%
4-Methyl-2-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NSn |
MDL Number:
MFCD07787403 CAS Number: 301652-23-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...