5-Methyl-2-(tributylstannyl)pyridine ≥94%

Supplier: Matrix Scientific

069508-500MG 069508-1G 069508-5G
101940-518EA 349.23 USD
101940-518 101940-520 101940-522
5-Methyl-2-(tributylstannyl)pyridine ≥94%
5-Methyl-2-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₃NSn
MDL Number: MFCD07787402
CAS Number: 189195-41-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR