2-Methyl-6-(tributylstannyl)pyridine ≥96%

Supplier: Matrix Scientific

069507-1G 069507-5G
101940-514EA 166.15 USD
101940-514 101940-516
2-Methyl-6-(tributylstannyl)pyridine ≥96%
2-Methyl-6-(tributylstannyl)pyridine

Hazard: Irritant

Formula: C₁₈H₃₃NSn
MW: 382.16
MDL Number: MFCD07787401
CAS Number: 259807-95-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR