4-Butylthiophenylboronic acid

Supplier: Matrix Scientific

Synonyms: (4-(Butylthio)phenyl)boronic acid

069505-1G 069505-5G 069505-25G
101940-500EA 133.85 USD
101940-500 101940-502 101940-504
4-Butylthiophenylboronic acid
4-Butylthiophenylboronic acid

Hazard: Irritant

Formula: C₁₀H₁₅BO₂S
MW: 210.1 g/mol
MDL Number: MFCD06801729
CAS Number: 151588-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR