4-(Cyclohexylcarbamoyl)phenylboronic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 4-(Cyclohexylaminocarbonyl)phenylboronic acid

069504-1G 069504-5G
101940-496EA 320 USD
101940-496 101940-498
4-(Cyclohexylcarbamoyl)phenylboronic acid ≥97%
4-(Cyclohexylcarbamoyl)phenylboronic acid

Hazard: Irritant

Formula: C₁₃H₁₈BNO₃
MDL Number: MFCD03411956
CAS Number: 762262-07-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR