2-Methoxy-3-(tributylstannyl)pyridine ≥95%
Supplier: Matrix Scientific
069503-1G
069503-5G
101940-492EA
349.23
USD
101940-492
101940-494
2-Methoxy-3-(tributylstannyl)pyridine ≥95%
2-Methoxy-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₈H₃₃NOSn |
MDL Number:
MFCD01319029 CAS Number: 223418-74-4 UN: 2788 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...