2-Fluoro-3-(tributylstannyl)pyridine ≥96%
Supplier: Matrix Scientific
069502-500MG
069502-1G
069502-5G
101940-486EA
349.23
USD
101940-486
101940-488
101940-490
2-Fluoro-3-(tributylstannyl)pyridine ≥96%
2-Fluoro-3-(tributylstannyl)pyridine
Hazard: Irritant
Formula:
C₁₇H₃₀FNSn |
CAS Number:
155533-81-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...