5-Cyano-2-thiopheneboronic acid ≥95%

Supplier: Matrix Scientific

Synonyms: (5-Cyanothiophen-2-yl)boronic acid, 5-Cyanothiophene-2-boronic acid

069497-1G 069497-5G
101940-458EA 72.31 USD
101940-458 101940-460
5-Cyano-2-thiopheneboronic acid ≥95%
5-Cyano-2-thiopheneboronic acid

Hazard: Irritant/Store in Freezer

Formula: C₅H₄BNO₂S
MW: 152.97 g/mol
MDL Number: MFCD02094029
CAS Number: 305832-67-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR