4-Iodo-6-methoxypyrimidine ≥98%

Supplier: Matrix Scientific

069494-1G 069494-5G 069494-25G
101940-440EA 198.46 USD
101940-440 101940-442 101940-444
4-Iodo-6-methoxypyrimidine ≥98%
4-Iodo-6-methoxypyrimidine

Hazard: Irritant

Formula: C₅H₅IN₂O
MW: 236.0100 g/mol
MDL Number: MFCD07787396
CAS Number: 161489-05-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR