cis-1-Ethoxy-2-(tributylstannyl)ethene ≥94%

Supplier: Matrix Scientific

069488-1G 069488-5G 069488-25G
101940-406EA 230.77 USD
101940-406 101940-408 101940-410
cis-1-Ethoxy-2-(tributylstannyl)ethene ≥94%
cis-1-Ethoxy-2-(tributylstannyl)ethene

Hazard: Toxic/Marine Pollutant

Formula: C₁₆H₃₄OSn
Boiling Pt: 84…87 °C
Density: 1.082
Flash Pt: >110 °C
MDL Number: MFCD07787391
CAS Number: 64724-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR