3-Bromo-1-(phenylsulfonyl)indole ≥97%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-1-(phenylsulphonyl)-1H-indole

069483-1G 069483-5G
101940-382EA 87.69 USD
101940-382 101940-384
3-Bromo-1-(phenylsulfonyl)indole ≥97%
3-Bromo-1-(phenylsulfonyl)indole

Hazard: Irritant

Formula: C₁₄H₁₀BrNO₂S
MW: 336.203 g/mol
MDL Number: MFCD02681986
CAS Number: 99655-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR