2,6-Dichloro-3-(tributylstannyl)pyrazine ≥96%

Supplier: Matrix Scientific

069482-500MG 069482-1G 069482-5G
101940-376EA 349.23 USD
101940-376 101940-378 101940-380
2,6-Dichloro-3-(tributylstannyl)pyrazine ≥96%
2,6-Dichloro-3-(tributylstannyl)pyrazine
Formula: C₁₆H₂₈Cl₂N₂Sn
MDL Number: MFCD07787382
CAS Number: 446285-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR