2-(N,N-Dimethylamino)pyridine-5-boronic acid dihydrochloride ≥96%

Supplier: Matrix Scientific

069480-1G 069480-5G
101940-366EA 295.38 USD
101940-366 101940-368
2-(N,N-Dimethylamino)pyridine-5-boronic acid dihydrochloride ≥96%
2-(N,N-Dimethylamino)pyridine-5-boronic acid dihydrochloride

Hazard: Irritant

Formula: C₇H₁₃BCl₂N₂O₂
MW: 238.91 g/mol
MDL Number: MFCD07189461
CAS Number: 265664-54-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR