2-(N-Boc)-5-(tributylstannyl)thiazole ≥96%

Supplier: Matrix Scientific

Synonyms: 2-Amino-5-(tributylstannyl)-1,3-thiazole, 2-BOC protected

069478-500MG 069478-1G 069478-5G
101940-354EA 756.92 USD
101940-354 101940-356 101940-358
2-(N-Boc)-5-(tributylstannyl)thiazole ≥96%
2-(N-Boc)-5-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₂₀H₃₈N₂O₂SSn
MDL Number: MFCD07787377
CAS Number: 243972-26-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR