4-Bromo-2-(thiomethyl)thiazole ≥98%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-(methylthio)thiazole

069477-1G 069477-5G 069477-25G
101940-348EA 320 USD
101940-348 101940-350 101940-352
4-Bromo-2-(thiomethyl)thiazole ≥98%
4-Bromo-2-(thiomethyl)thiazole

Hazard: Irritant

Formula: C₄H₄BrNS₂
MW: 210.12 g/mol
MDL Number: MFCD07787376
CAS Number: 204513-62-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR