2-Ethoxy-4-(tributylstannyl)thiazole ≥96%

Supplier: Matrix Scientific

069474-500MG 069474-1G 069474-5G
101940-330EA 553.85 USD
101940-330 101940-332 101940-334
2-Ethoxy-4-(tributylstannyl)thiazole ≥96%
2-Ethoxy-4-(tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₇H₃₃NOSSn
MW: 418.21
MDL Number: MFCD07787373
CAS Number: 240816-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR