2-(Methylthio)-5-pyrimidineboronic acid ≥97%
Supplier: Matrix Scientific
Synonyms:
(2-(Methylthio)pyrimidin-5-yl)boronic acid, 2-(Methylthio)pyrimidine-5-boronic acid
069472-1G
069472-5G
069472-25G
101940-318EA
344.62
USD
101940-318
101940-320
101940-322
2-(Methylthio)-5-pyrimidineboronic acid ≥97%
2-(Methylthio)-5-pyrimidineboronic acid
Hazard: Irritant
Formula:
C₅H₇BN₂O₂S MW: 170.0000 g/mol |
MDL Number:
MFCD07375142 CAS Number: 348098-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...