2-(Methylthio)-5-(tributylstannyl)pyrimidine ≥95%

Supplier: Matrix Scientific

069471-500MG 069471-1G 069471-5G
101940-312EA 524.62 USD
101940-312 101940-314 101940-316
2-(Methylthio)-5-(tributylstannyl)pyrimidine ≥95%
2-(Methylthio)-5-(tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₇H₃₂N₂SSn
MDL Number: MFCD07787371
CAS Number: 120717-37-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR