2-(Methylthio)-4-(tributylstannyl)pyrimidine ≥96%

Supplier: Matrix Scientific

069470-500MG 069470-1G 069470-5G
101940-306EA 233.85 USD
101940-306 101940-308 101940-310
2-(Methylthio)-4-(tributylstannyl)pyrimidine ≥96%
2-(Methylthio)-4-(tributylstannyl)pyrimidine

Hazard: Irritant

Formula: C₁₇H₃₂N₂SSn
MDL Number: MFCD01319026
CAS Number: 123061-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR