4-(Tributylstannyl)thiazole ≥94%
Supplier: Matrix Scientific
069469-500MG
069469-1G
069469-5G
101940-300EA
184.62
USD
101940-300
101940-302
101940-304
4-(Tributylstannyl)thiazole ≥94%
4-(Tributylstannyl)thiazole
Hazard: Toxic
Formula:
C₁₅H₂₉NSSn MW: 374.17 |
MDL Number:
MFCD01319073 CAS Number: 173979-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...