5-(Tributylstannyl)thiazole ≥93%

Supplier: Matrix Scientific

069468-500MG 069468-1G 069468-5G
101940-294EA 349.23 USD
101940-294 101940-296 101940-298
5-(Tributylstannyl)thiazole ≥93%
5-(Tributylstannyl)thiazole

Hazard: Irritant

Formula: C₁₅H₂₉NSSn
MDL Number: MFCD01319074
CAS Number: 157025-33-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR